missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat IMPA1/IMP1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB98901-SP
This item is not returnable.
View return policy
Description
IMPA1 Monoclonal specifically detects IMPA1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| IMPA1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| EC 3.1.3.25, IMP, IMP 1, IMP1, IMPA, IMPase 1, inositol monophosphatase 1, inositol(myo)-1(or 4)-monophosphatase 1, Inositol-1(or 4)-monophosphatase 1, Lithium-sensitive myo-inositol monophosphatase A1, MRT59, Myo-Inositol Monophosphatase 1 | |
| Synthetic peptide containing human IMPA/IMPA1 Peptide Accession # P29218 | |
| 25 μg | |
| 3612 | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 984603 | |
| Western Blot 1μg/mL | |
| P29218 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human IMPA/IMPA1 Peptide in direct ELISAs. Detects human, mouse, and rat IMPA/IMPA1 in Western blots. | |
| Human, Mouse, Rat | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction