missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat ILK Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4266
This item is not returnable.
View return policy
Description
ILK Monoclonal specifically detects ILK in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| ILK | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q13418 | |
| ILK | |
| E. coli-derived recombinant human ILK Met1-Lys452 Accession # Q13418 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat ILK in Western blots. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| 443208 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 59 kDa serine/threonine-protein kinase, DKFZp686F1765, EC 2.7.11.1, ESTM24, ILK1, ILK-1, ILK2, ILK-2, integrin-linked kinase, integrin-linked kinase-2, integrin-linked protein kinase, p59, p59ILK | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3611 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction