missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat HTRA2/Omi Antibody, R&D Systems™
Rabbit Polyclonal Antibody has been used in 22 publications
Brand: R&D Systems AF1458
This item is not returnable.
View return policy
Description
HTRA2/Omi Polyclonal specifically detects HTRA2/Omi in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.
Specifications
| HTRA2/Omi | |
| Polyclonal | |
| Unconjugated | |
| O43464 | |
| HTRA2 | |
| E. coli-derived recombinant human HTRA2/Omi Ala134-Glu458 Accession # O43464 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat full length and mitochondria-processed HTRA2/Omi. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.25 ug/mL, Simple Western 2.5 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| High temperature requirement protein A2, HtrA serine peptidase 2, HtrA2, HtrA-like serine protease, Omi, OMIOmi stress-regulated endoprotease, PARK13EC 3.4.21.108, PRSS25, PRSS25serine, 25, Serine protease 25, serine protease HTRA2, mitochondrial, Serine proteinase OMI | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 27429 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction