missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat HSPH1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF4029
This item is not returnable.
View return policy
Description
HSPH1/HSP105 Polyclonal specifically detects HSPH1/HSP105 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| HSPH1/HSP105 | |
| Polyclonal | |
| Unconjugated | |
| Q66HA8 | |
| HSPH1 | |
| E. coli-derived recombinant rat HSPH1 Val595-Asp733 Accession # Q66HA8 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat HSPH1. This antibody is predicted to react with HSPH1 alpha and beta isoforms based on sequence identity of the immunogen. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Antigen NY-CO-25, heat shock 105kD beta, heat shock 105kDa protein 1, heat shock 105kDa/110kDa protein 1, Heat shock 110 kDa protein, heat shock protein 105 kDa, HSP105a, HSP105B, HSP105heat shock 105kD alpha, HSP110, KIAA0201DKFZp686M05240, NY-CO-25 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 10808 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction