missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat HSP90 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF3775
This item is not returnable.
View return policy
Description
HSP90 Polyclonal specifically detects HSP90 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.
Specifications
| HSP90 | |
| Polyclonal | |
| Unconjugated | |
| P07900 | |
| HSP90AA1 | |
| E. coli-derived recombinant human HSP90 alpha Met1-Asp732 Accession # P07900 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat HSP90 in Western blots. Detects recombinant human HSP90 alpha and HS90 beta in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| D6S182, FLJ31884, Heat shock 86 kDa, heat shock 90kD protein 1, alpha, heat shock 90kD protein 1, alpha-like 4, heat shock 90kD protein, alpha-like 4, heat shock 90kDa protein 1, alpha, heat shock protein 90kDa alpha (cytosolic), class A member 1, heat shock protein HSP 90-alpha, HSP84, HSP86, Hsp89, HSP89A, Hsp90, HSP90A, HSP90AA1, HSP90AB1, HSP90AHSP86, HSP90B, HSP90N, HSPC1, HSPC1HSP 86, HSPC2, HSPCA, HSPCAHSP89A, HSPCAL1, HSPCAL4, HSPCB, HSPN, LAP2, Renal carcinoma antigen NY-REN-38 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3320 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction