missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat HSP70/HSPA1A Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
£415.00
Specifications
| Antigen | HSP70/HSPA1A |
|---|---|
| Clone | 242707 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
HSP70/HSPA1A Monoclonal specifically detects HSP70/HSPA1A in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
| HSP70/HSPA1A | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| 3303 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant |
| 242707 | |
| Monoclonal | |
| Purified | |
| RUO | |
| dnaK-type molecular chaperone HSP70-1, FLJ54303, FLJ54370, FLJ54392, FLJ54408, FLJ75127, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1A, heat shock 70kDa protein 1A, heat shock-induced protein, HSP70.1/HSP70.2, HSP70-1, HSP70-1/HSP70-2, HSP70-1A, HSP70I, HSP72, HSPA1, HSPA1A, HSPA1B | |
| HSPA1A | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title