missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat HSP60 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 6 publications
£430.00
Specifications
| Antigen | HSP60 |
|---|---|
| Clone | 264233 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Beschreibung
HSP60 Monoclonal specifically detects HSP60 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Spezifikation
| HSP60 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 60 kDa chaperonin, Chaperonin 60, CPN60, GROEL, heat shock 60kD protein 1 (chaperonin), heat shock 60kDa protein 1 (chaperonin), Heat shock protein 60, heat shock protein 65, HLD4, Hsp60, HSP-60, HSP60SPG13, HSP65, HSPD1, HuCHA60, Mitochondrial matrix protein P1,60 kDa heat shock protein, mitochondrial, P60 lymphocyte protein, short heat shock protein 60 Hsp60s1, spastic paraplegia 13 (autosomal dominant), SPG13 | |
| HSPD1 | |
| Primary |
| 264233 | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| 3329 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts