missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse/Rat HNF-3 beta/FoxA2 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18709878
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18709878 100 μg 100µg
18794168 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18709878 Supplier R&D Systems Supplier No. MAB2400100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

HNF-3 beta/FoxA2 Monoclonal specifically detects HNF-3 beta/FoxA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HNF-3 beta/FoxA2
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 2141A
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 3-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q9Y261
Gene Alias forkhead box A2, Forkhead box protein A2, FoxA2, hepatic nuclear factor-3-beta, hepatocyte nuclear factor 3, beta, hepatocyte nuclear factor 3-beta, HNF3 beta, HNF-3B, HNF-3-beta, HNF3BTCF-3B, MGC19807, TCF3B, Transcription factor 3B
Host Species Rabbit
Immunogen Human/Mouse/Rat HNF-3 beta/FoxA2 synthetic peptide Accession # Q9Y261
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3170
Test Specificity Detects human HNF-3 beta/FoxA2 in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.