missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Histamine H3 R Antibody, R&D Systems™

Rabbit Monoclonal Antibody
£166.00 - £412.00
Specifications
| Antigen | Histamine H3R |
|---|---|
| Clone | 2420B |
| Dilution | Flow Cytometry 0.25 μg/106 cells, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18702897
![]() |
R&D Systems
MAB102001-SP |
25 μg |
£166.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18742037
![]() |
R&D Systems
MAB102001-100 |
100 μg |
£412.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Histamine H3R Monoclonal specifically detects Histamine H3R in Human, Mouse, Rat samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| Histamine H3R | |
| Flow Cytometry 0.25 μg/106 cells, CyTOF-ready | |
| Monoclonal | |
| Rabbit | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| GPCR97, GPCR97G protein-coupled receptor 97, G-protein coupled receptor 97, H3R, HH3R, histamine H3 receptor, Histamine H3R, histamine receptor H3, HRH3 | |
| Synthetic peptide containing Human/Mouse/Rat Histamine H3 R, Accession # Q9Y5N1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human, mouse, and rat Histamine H3 R in direct ELISAs. |
| 2420B | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Human, Mouse, Rat | |
| Q9Y5N1 | |
| 11255 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title