missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat HIF-2 alpha/EPAS1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 15 publications
Brand: R&D Systems AF2997
This item is not returnable.
View return policy
Description
HIF-2 alpha/EPAS1 Polyclonal specifically detects HIF-2 alpha/EPAS1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| HIF-2 alpha/EPAS1 | |
| Polyclonal | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Basic-helix-loop-helix-PAS protein MOP2, BHLHE73, Class E basic helix-loop-helix protein 73, ECYT4, endothelial PAS domain protein 1, endothelial PAS domain-containing protein 1, EPAS1, EPAS-1, HIF 2A, HIF-1-alpha-like factor, HIF-1alpha-like factor, HIF2 alpha, HIF-2 alpha, HIF2A, hif2a angiogenesis, HIF-2-alpha, HIF2-alpha, HLF, hypoxia-inducible factor 2 alpha, Hypoxia-inducible factor 2-alpha, Member of PAS protein 2, MOP2, PAS domain-containing protein 2, PASD2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2034 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| AAH57870 | |
| EPAS1 | |
| E. coli-derived recombinant mouse HIF-2 alpha/EPAS1 Ser542-Thr874 Accession # AAH57870 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat HIF-2 alpha/EPAS1 in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction