missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Hepassocin/FGL1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1614-SP
This item is not returnable.
View return policy
Description
Hepassocin/FGL1 Polyclonal specifically detects Hepassocin/FGL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry.
Specifications
| Hepassocin/FGL1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FGL1, fibrinogen-like 1, fibrinogen-like protein 1, Fibrinogen-like-protein 1, FREP1, Hepassocin, hepatocellular carcinoma-related sequence, Hepatocyte-derived fibrinogen-related protein 1, HFREP-1, HFREP1MGC12455, HP-041, LFIRE1, LFIRE-1, Liver fibrinogen-related protein 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2267 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| BAB70690 | |
| FGL1 | |
| Mouse myeloma cell line NS0-derived recombinant human Hepassocin Met1-Ile312 Accession # BAB70690 | |
| 25 μg | |
| Primary | |
| Detects human Hepassocin in direct ELISAs and Western blots. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction