missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse/Rat GRK2 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18732129
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18732129 25 μg 25µg
18776049 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18732129 Supplier R&D Systems Supplier No. MAB43391SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

GRK2 Monoclonal specifically detects GRK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Knockout Validated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRK2
Applications Western Blot, Immunoassay
Classification Monoclonal
Clone 2089B
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Knockout Validated
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
Gene Accession No. P25098
Gene Alias ADRBK1, adrenergic, beta, receptor kinase 1, BARK, BARK1, BARK1beta-ARK-1, Beta-ARK-1, BETA-ARK1, EC 2.7.11, EC 2.7.11.15, FLJ16718, G-protein coupled receptor kinase 2, GRK2beta-adrenergic receptor kinase 1
Host Species Rabbit
Immunogen E. coli-derived recombinant human GRK2 Pro468-Leu689 Accession # P25098
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 156
Test Specificity Detects human GRK2 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.