missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat GRK2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4339
This item is not returnable.
View return policy
Description
GRK2 Monoclonal specifically detects GRK2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| GRK2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P25098 | |
| ADRBK1 | |
| E. coli-derived recombinant human GRK2 Pro468-Leu689 Accession # P25098 | |
| 100 μg | |
| Primary | |
| Detects endogenous human, mouse and rat GRK2 in Western blots. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| 466717 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ADRBK1, adrenergic, beta, receptor kinase 1, BARK, BARK1, BARK1beta-ARK-1, Beta-ARK-1, BETA-ARK1, EC 2.7.11, EC 2.7.11.15, FLJ16718, G-protein coupled receptor kinase 2, GRK2beta-adrenergic receptor kinase 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 156 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction