missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Galectin-4 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1227-SP
This item is not returnable.
View return policy
Description
Galectin-4 Polyclonal specifically detects Galectin-4 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| Galectin-4 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Antigen NY-CO-27, gal-4, GAL4, galectin 4, galectin-4, Galectin4, L-36 lactose-binding protein, L36LBP, Lactose-binding lectin 4, lectin, galactoside-binding, soluble, 4, LGALS4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3960 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| P56470 | |
| LGALS4 | |
| E. coli-derived recombinant human Galectin-4 Ala2-Ile323 Accession # P56470 | |
| 25 μg | |
| Primary | |
| Detects human Galectin-4 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) Galectin-2 is observed, and less than 1% cross-reactivity with rhGalectin-1, rhGalectin-3, rhGalectin-7, and rhGalectin-8 is observed. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction