missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat FBPase 1 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF7285
This item is not returnable.
View return policy
Description
FBPase 1 Polyclonal specifically detects FBPase 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| FBPase 1 | |
| Polyclonal | |
| Unconjugated | |
| P09467 | |
| FBP1 | |
| E. coli-derived recombinant human FBPase 1 Ala2-Gln338 (Arg218Lys) Accession # P09467 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, rat FBPase 1 in Western blots and detects human FBPase 1 in direct ELISAs. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.2 ug/mL | |
| D-fructose-1,6-bisphosphate 1-phosphohydrolase 1, EC 3.1.3, EC 3.1.3.11, FBP, FBP1, FBPase 1, fructose-1,6-bisphosphatase 1, fructose-bisphosphatase 1, growth-inhibiting protein 17 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 2203 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction