missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat FABP5/E-FABP Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 9 publications
Brand: R&D Systems AF1476-SP
This item is not returnable.
View return policy
Description
FABP5/E-FABP Polyclonal specifically detects FABP5/E-FABP in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Knockout Validated.
Specifications
| FABP5/E-FABP | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EFABP, E-FABP, E-FABPPAFABP, fatty acid binding protein 5 (psoriasis-associated), Fatty acid-binding protein 5, PA-FABPepidermal, Psoriasis-associated fatty acid-binding protein homolog | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2171 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Immunoassay | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 3-15 ug/mL, Knockout Validated | |
| Q05816 | |
| FABP5 | |
| E. coli-derived recombinant mouse FABP5/E-FABP Met1-Gln135 Accession # Q05816 | |
| 25 μg | |
| Primary | |
| Detects mouse FABP5/E-FABP in direct ELISAs and Western blots. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction