missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat ERK1 Antibody, R&D Systems™
Rabbit Polyclonal Antibody
Brand: R&D Systems AF1575
This item is not returnable.
View return policy
Description
ERK1 Polyclonal specifically detects ERK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| ERK1 | |
| Polyclonal | |
| Unconjugated | |
| XM_055766 | |
| MAPK3 | |
| E. coli-derived recombinant human ERK1 Accession # XM_055766 | |
| 50 μg | |
| Primary | |
| Detects human, mouse, and rat ERK1 in Western blots. Detects neither recombinant nor endogenous ERK2 in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| EC 2.7.11, ERK-1, ERK1p44-MAPK, ERT2, Extracellular signal-regulated kinase 1, extracellular signal-related kinase 1, HS44KDAP, HUMKER1A, Insulin-stimulated MAP2 kinase, MAP kinase 1, MAP kinase 3, MAP kinase isoform p44, MAPK 1, MAPK 3, MAPK3, MGC20180, Microtubule-associated protein 2 kinase, Mitogen-activated protein kinase 1, mitogen-activated protein kinase 3, p44erk1, p44-ERK1, P44MAPK, PRKM3, PRKM3EC 2.7.11.24 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 5595 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction