missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse/Rat DDX1 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18797999
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18797999 25 μg 25µg
18789657 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18797999 Supplier R&D Systems Supplier No. MAB9615SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

DDX1 Monoclonal specifically detects DDX1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 2220A
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 1-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q92499
Gene Alias ATP-dependent RNA helicase DDX1, DBP-RB, DBP-RBDEAD box polypeptide 1, DEAD (Asp-Glu-Ala-Asp) box polypeptide 1, DEAD box protein 1, DEAD box protein retinoblastoma, DEAD box-1, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 1, DEAD/H-Box Helicase 1, EC 3.6.1, EC 3.6.1.10, EC 3.6.4.13, UKVH5d
Host Species Rabbit
Immunogen Human/mouse/rat DDX1 synthetic Peptide Accession # Q92499
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 1653
Test Specificity Detects human, mouse, and rat DDX1 in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.