missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat CUGBP1/CELF1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£157.00 - £370.00
Specifications
| Antigen | CUGBP1/CELF1 |
|---|---|
| Clone | 850717 |
| Dilution | Western Blot 0.5 ug/mL, Immunohistochemistry 8-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18718538
![]() |
R&D Systems
MAB9388-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18795569
![]() |
R&D Systems
MAB9388-100 |
100 μg |
£370.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CUGBP1/CELF1 Monoclonal specifically detects CUGBP1/CELF1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| CUGBP1/CELF1 | |
| Western Blot 0.5 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human, Mouse, Rat | |
| Q92879 | |
| 10658 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 850717 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| BRUNOL2, BRUNOL250 kDa nuclear polyadenylated RNA-binding protein, bruno-like 2, Bruno-like protein 2, CELF1, CUG RNA-binding protein, CUG triplet repeat, RNA binding protein 1, CUG triplet repeat, RNA-binding protein 1, CUG-BP, CUG-BP- and ETR-3-like factor 1, CUGBP Elav-like family member 1, CUGBP, Elav-like family member 1, CUG-BP1, CUGBP1CUG triplet repeat RNA-binding protein 1, CUGBPCELF-1, D2Wsu101e, Deadenylation factor CUG-BP, EDEN-BP, EDEN-BP homolog, embryo deadenylation element binding protein, Embryo deadenylation element-binding protein homolog, hNab50, NAB50, NAB50CUG-BP1, NAPOR, nuclear polyadenylated RNA-binding protein, 50-kD, RNA-binding protein BRUNOL-2 | |
| E. coli-derived recombinant human CUGBP1/CELF1 Met1-Gly60 Accession # Q92879 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CUGBP1/CELF1 in direct ELISAs and Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title