missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Complexin-2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF5085
This item is not returnable.
View return policy
Description
Complexin-2 Polyclonal specifically detects Complexin-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Complexin-2 | |
| Polyclonal | |
| Unconjugated | |
| Q6PUV4 | |
| CPLX2 | |
| E. coli-derived recombinant human Complexin-2 Met1-Lys134 Accession # Q6PUV4 | |
| 100 μg | |
| Primary | |
| Detects endogenous human, mouse and rat Complexin-2 in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| complexin 2, Complexin II, complexin-2, CPLX2, CPX II, CPX2, CPX-2, CPX-II, DKFZp547D155, Hfb1, MGC138492, Synaphin 1, synaphin 1,921-L, synaphin-1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 10814 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction