missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Complexin-1/2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8069-SP
This item is not returnable.
View return policy
Description
Complexin-1/2 Monoclonal specifically detects Complexin-1/2 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| Complexin-1/2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| O14810 | |
| E. coli-derived recombinant human Complexin-1 Met1-Lys134 Accession # O14810 | |
| 25 μg | |
| Primary | |
| Detects human Complexin-1 and Complexin-2 in ELISAs. Detects human, mouse, and rat Complexin-1 and -2 in Western blots. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 837802 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10815 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction