missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Caveolin-2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF5788
This item is not returnable.
View return policy
Description
Caveolin-2 Polyclonal specifically detects Caveolin-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Specifications
| Caveolin-2 | |
| Polyclonal | |
| Unconjugated | |
| P51636 | |
| CAV2 | |
| E. coli-derived recombinant human Caveolin-2 Met1-Lys86 Accession # P51636 | |
| 100 μg | |
| Primary | |
| Detects endogenous human, mouse, and rat Caveolin-2 in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| CAV, CAV2, caveolae protein, 20-kD, caveolin 2, caveolin 2 isoform a and b, caveolin-2, Caveolin2, MGC12294 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 858 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction