missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Casein Kinase 1 gamma Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF3186
This item is not returnable.
View return policy
Description
Casein Kinase 1 gamma Polyclonal specifically detects Casein Kinase 1 gamma in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Casein Kinase 1 gamma | |
| Polyclonal | |
| Unconjugated | |
| Q9HCP0 | |
| CSNK1G1 | |
| E. coli-derived recombinant human CK1 gamma Val182-Tyr304 Accession # Q9HCP0 | |
| 100 μg | |
| Primary | |
| Detects endogenous human, mouse, and rat CK1 gamma in Western blots. In Western blots, the immunogen is derived from a region of human CK1 gamma that is 97% to 98% identical to human CK1 gamma 2 and CK1 gamma 3 so all three isoforms should be detected. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 3-15 ug/mL | |
| casein kinase 1, gamma 1, casein kinase I isoform gamma-1, CKI-gamma 1, CSNK1G1, EC 2.7.11, EC 2.7.11.1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 53944 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction