missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat BNIP3L Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF4030-SP
This item is not returnable.
View return policy
Description
BNIP3L Polyclonal specifically detects BNIP3L in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| BNIP3L | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Adenovirus E1B19K-binding protein B5, BCL2/adenovirus E1B 19 kDa protein-interacting protein 3A, BCL2/adenovirus E1B 19 kDa protein-interacting protein 3-like, BCL2/adenovirus E1B 19-kd protein-interacting protein 3a, BCL2/adenovirus E1B 19kDa interacting protein 3-like, BNIP3a, BNIP3H, Nip3L, NIP3-like protein X, Nix, NIXBCL2/adenovirus E1B 19kD-interacting protein 3-like | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 665 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| O60238 | |
| BNIP3L | |
| E. coli-derived recombinant human BNIP3L Ser2-Glu184 Accession # O60238 | |
| 25 μg | |
| Primary | |
| Detects endogenous human, mouse and rat BNIP3L in Western blots. In Western blots, less than 10% cross-reactivity to recombinant human BNIP3L is observed. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction