missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Biliverdin Reductase B/BLVRB Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6568
This item is not returnable.
View return policy
Description
Biliverdin Reductase B/BLVRB Polyclonal specifically detects Biliverdin Reductase B/BLVRB in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Biliverdin Reductase B/BLVRB | |
| Polyclonal | |
| Unconjugated | |
| P30043 | |
| BLVRB | |
| E. coli-derived recombinant human Biliverdin Reductase B/BLVRB Ala2-Gln206, predicted Accession # P30043 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat Biliverdin Reductase B/BLVRB in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Biliverdin reductase B, biliverdin reductase B (flavin reductase (NADPH)), Biliverdin-IX beta-reductase, BLVRB, BVRB, BVR-B, EC 1.3.1.24, EC 1.5.1.30, flavin reductase, flavin reductase (NADPH), FLR, FLRBVRB, FR, GHBP, Green heme-binding protein, MGC117413, NADPH-dependent diaphorase, NADPH-flavin reductase, SDR43U1, short chain dehydrogenase/reductase family 43U, member 1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 645 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction