missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat beta-Catenin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 16 publications
Brand: R&D Systems AF1329-SP
This item is not returnable.
View return policy
Description
beta-Catenin Polyclonal specifically detects beta-Catenin in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry, CyTOF-ready.
Specifications
| beta-Catenin | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| bCatenin, beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, CTNNB1, DKFZp686D02253, FLJ25606, FLJ37923 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1499 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| ChIP Assay, CyTOF, Flow Cytometry, Immunocytochemistry, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 50 ug/mL, Immunohistochemistry 5-15 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Chromatin Immunoprecipitation (ChIP) 5 ug/5 x 10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
| P35222 | |
| CTNNB1 | |
| E. coli-derived recombinant human beta-Catenin Ala2-Leu781 Accession # P35222 | |
| 25 μg | |
| Primary | |
| Detects human, mouse and rat beta-Catenin in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction