missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat ARHGAP1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6407-SP
This item is not returnable.
View return policy
Description
ARHGAP1 Polyclonal specifically detects ARHGAP1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| ARHGAP1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CDC42 GTPase-activating protein, CDC42GAP, CDC42GAPp50-RhoGAP, GTPase-activating protein rhoOGAP, p50rhoGAP, Rho GTPase activating protein 1, rho GTPase-activating protein 1, RHOGAP, RHOGAP1Cdc42GAP, Rho-related small GTPase protein activator, Rho-type GTPase-activating protein 1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 392 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.2 ug/mL | |
| Q07960 | |
| ARHGAP1 | |
| E. coli-derived recombinant human ARHGAP1 aa 1-62 Accession # Q07960 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat ARHGAP1 in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction