missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat APE Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB1044
This item is not returnable.
View return policy
Description
APE Monoclonal specifically detects APE in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| APE | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| P27695 | |
| APEX1 | |
| E. coli-derived recombinant human APE Pro2-Leu318 Accession # P27695 | |
| 100 μg | |
| Primary | |
| Detects human, mouse and rat APE in Western blots. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| 200913 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AP endonuclease 1, APE, APE1, APE-1, APEAPEX nuclease (multifunctional DNA repair enzyme), APENAP endonuclease class I, APEX deoxyribonuclease (apurinic or apyrimidinic), APEX nuclease, APEX nuclease (multifunctional DNA repair enzyme) 1, APEX1, Apurinic-apyrimidinic endonuclease 1, APXAP lyase, DNA-(apurinic or apyrimidinic site) lyase, EC 3.1, EC 4.2.99.18, HAP1apurinic/apyrimidinic (abasic) endonuclease, multifunctional DNA repair enzyme, protein REF-1, redox factor 1, Redox factor-1, REF-1, REF1 apurinic/apyrimidinic exonuclease | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 328 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction