missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat APBA2 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF6327
This item is not returnable.
View return policy
Description
APBA2 Polyclonal specifically detects APBA2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| APBA2 | |
| Polyclonal | |
| Unconjugated | |
| Q99767 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 321 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| APBA2 | |
| E. coli-derived recombinant human APBA2 Ala2-Leu165 Accession # Q99767 | |
| 100 μg | |
| Primary | |
| Detects human APBA2 in direct ELISAs and human, mouse and rat in Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) APBA1 and rhAPBA3 is observed. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction