missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Annexin A2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3928
This item is not returnable.
View return policy
Description
Annexin A2 Monoclonal specifically detects Annexin A2 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| Annexin A2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P07355 | |
| ANXA2 | |
| E. coli-derived recombinant human Annexin A2 Met1-Asp339 Accession # P07355 | |
| 100 μg | |
| Primary | |
| Detects human Annexin A2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 25% cross-reactivity with recombinant human (rh) Annexin A1, A7, A9, A11, and A13 is observed. No cross-reactivity with rhAnnexin A3, A4, A6, A8, or A10 is observed. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 666316 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| annexin A2, Annexin II, annexin-2, ANX2, ANX2L4, ANX2L4LPC2, ANXA2, CAL1H, Calpactin I heavy chain, calpactin I heavy polypeptide, Calpactin-1 heavy chain, chromobindin 8, chromobindin-8, LIP2, LIP2PAP-IV, Lipocortin II, Lipocortin-2, LPC2D, LPC2DP36, p36, PAP-IV, Placental anticoagulant protein IV, Protein I | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 302 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction