missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat ALDH2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB10168-SP
This item is not returnable.
View return policy
Description
ALDH2 Monoclonal specifically detects ALDH2 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.
Specifications
| ALDH2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| acetaldehyde dehydrogenase 2, aldehyde dehydrogenase 2 family (mitochondrial), aldehyde dehydrogenase, mitochondrial, ALDH class 2, ALDH-E2, ALDHI, ALDM, EC 1.2.1, EC 1.2.1.3, liver mitochondrial ALDH, MGC1806, nucleus-encoded mitochondrial aldehyde dehydrogenase 2 | |
| Synthetic peptide containing human ALDH2, Ser18-Ser517, Accession # P05091 | |
| 25 μg | |
| 217 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| 1007902 | |
| Western Blot 2 μg/mL, Simple Western 20 μg/mL, Immunohistochemistry 0.5-25 μg/mL, Immunocytochemistry 8-25 μg/mL | |
| P05091 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human ALDH2 in direct ELISAs. Detects human, mouse, and rat ALDH2 in Western blots. | |
| Human, Mouse, Rat | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction