missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Akt Pan Specific PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC2055P
This item is not returnable.
View return policy
Description
AKT Monoclonal specifically detects AKT in Human, Mouse, Rat samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| AKT | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| RAC-ALPHA, AKT, AKT serine/threonine kinase 1, PKB, PKB alpha, PKB beta, PKB gamma, PKB-ALPHA, PRKBA, RAC, RAC-alpha, RAC-beta, RAC-gamma | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 207 | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| 281046 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| P31749 | |
| AKT1 | |
| E. coli-derived recombinant human Akt1 Ser2-Ala480 Accession # P31749 | |
| 100 Tests | |
| Primary | |
| Detects human, mouse and rat Akt in direct ELISAs and Western blots. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido