missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat AIF Biotinylated Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Brand: R&D Systems BAF1457
This item is not returnable.
View return policy
Description
AIF Polyclonal specifically detects AIF in Human, Mouse, Rat samples. It is validated for Immunocytochemistry.
Specifications
| AIF | |
| Polyclonal | |
| Biotin | |
| Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative | |
| AIFapoptosis-inducing factor 1, mitochondrial, AIFM1, apoptosis-inducing factor, mitochondrion-associated, 1, COXPD6, MGC111425, PDCD8, PDCD8striatal apoptosis-inducing factor, programmed cell death 8 (apoptosis-inducing factor), Programmed cell death protein 8 | |
| Rabbit | |
| Antigen Affinity-purified | |
| Primary | |
| AIF antibodies are ideal for immunocytochemistry colocalization studies in mitochondria. The unconjugated antibody detects human, mouse, and rat mitochondria-processed AIF. | |
| Human, Mouse, Rat | |
| Purified |
| Immunocytochemistry | |
| LYOPH | |
| Immunocytochemistry 5-15 ug/mL | |
| O95831 | |
| AIFM1 | |
| E. coli-derived recombinant human AIF Glu121-Asp613 Accession # O95831 | |
| 50 μg | |
| 9131 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction