missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human/Mouse/Rat Activin A Recombinant Protein, PeproTech®

Product Code. 30088404
Click to view available options
Quantity:
10 μg
100 μg
2 μg
2 x 500 μg
250 μg
50 μg
500 μg
missing translation for 'unitSize'
100µg
10µg
1mg
250µg
2µg
500µg
50µg
Pack of 2
This item is not returnable. View return policy

Product Code. 30088404

missing translation for 'mfr': Gibco™ 1201401M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

120-14-1MG will be provided as 2 x 500 μg (120-14-500UG). Recombinant Human/Murine/Rat Activin A is a 26.0 kDa disulfide-linked homodimer of two betaA chains, each containing 116 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Activins are dimeric growth and differentiation factors which belong to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. Activins signal through a heteromeric complex of receptor serine kinases which include at least two type I ( I and IB) and two type II (II and IIB) receptors. These receptors are all transmembrane proteins, composed of a ligand-binding extracellular domain with cysteine-rich region, a transmembrane domain, and a cytoplasmic domain with predicted serine/threonine specificity. Type I receptors are essential for signaling; and type II receptors are required for binding ligands and for expression of type I receptors. Type I and II receptors form a stable complex after ligand binding, resulting in phosphorylation of type I receptors by type II receptors. Activin A mutations are associated with fibrodysplasia ossificans progressive.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08476, P18331, Q04998
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 16323, 29200, 3624
Molecular Weight (g/mol) 26 kDa
Name Human/Mouse/Rat Activin A
Purification Method Purified
Quantity 2 x 500 μg
Source Insect cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias activin A; activin beta-A chain; BMP7; BMP-7; Bone morphogenetic protein 7; EDF; Eptotermin alfa; erythroid differentiation factor; erythroid differentiation protein; follicle-stimulating hormone-releasing protein; FRP; FSH-releasing protein; INHB; Inhba; inhibin B subunit; inhibin beta A chain; inhibin beta A protein; inhibin beta A subunit; inhibin beta-A; inhibin beta-A-subunit; inhibin beta-subunit; inhibin subunit beta A; inhibin, beta A; inhibin, beta A (activin A, activin AB alpha polypeptide); Inhibin, beta-1; OP1; OP-1; Osteogenic protein 1; unnamed protein product
Common Name Activin A
Gene Symbol Inhba
Biological Activity Determined by its ability to inhibit the proliferation of mouse MPC-11 cells. The expected ED50 is ≤ 2.0 ng/ml, corresponding to a specific activity of ≥ 5 x 10^5 units/mg
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence GLECDGKVNI CCKKQFFVSF KDIGWNDWII APSGYHANYC EGECPSHIAG TSGSSLSFHS TVINHYRMRG HSPFANLKSC CVPTKLRPMS MLYYDDGQNI IKKDIQNMIV EECGCS
Content And Storage -20°C
Activity Determined by its ability to inhibit the proliferation of mouse MPC-11 cells. The expected ED50 is ≤ 2.0 ng/ml, corresponding to a specific activity of ≥ 5 x 10^5 units/mg.
Expression System Insect cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.