missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse R-Spondin 3 Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 1 publication
Brand: R&D Systems MAB41201
Additional Details : Weight : 0.00970kg
Description
R-Spondin 3 Monoclonal specifically detects R-Spondin 3 in Human, Mouse samples. It is validated for Western Blot.Specifications
R-Spondin 3 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
BAC36296 | |
RSPO3 | |
E. coli-derived recombinant mouse R-Spondin 3 Ser21-Gly209 (Gln108Lys) Accession # BAC36296 | |
100 μg | |
Primary | |
Detects human and mouse R-Spondin 3 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) R-Spondin 1, recombinant human R-Spondin 2, or rmR-Spondin 4 is observed. | |
Human, Mouse | |
Purified |
Western Blot | |
400403 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Cristin 1, CRISTIN1, FLJ14440, hPWTSR, hRspo3, Protein with TSP type-1 repeat, PWTSR, Roof plate-specific spondin-3, RSPO3, RSpondin 3, R-spondin 3 homolog (Xenopus laevis), R-spondin-3, Thrombospondin type-1 domain-containing protein 2, thrombospondin, type I, domain containing 2, THSD2 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
84870 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
For Research Use Only