missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Primate BMP-9 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 11 publications
£181.00 - £1750.00
Specifications
| Antigen | BMP-9 |
|---|---|
| Clone | 360107 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Neutralization 0.01-0.04 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, Neutralization, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18721708
![]() |
R&D Systems
MAB3209-SP |
25 μg |
£181.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18131461
![]() |
R&D Systems
MAB3209-100 |
100 μg |
£429.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18150341
|
R&D Systems
MAB3209-500 |
500 μg |
£1750.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BMP-9 Monoclonal specifically detects BMP-9 in Human, Mouse, Primate samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
| BMP-9 | |
| LYOPH | |
| Western Blot, Neutralization, ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Primate | |
| Q9UK05 | |
| 2658 | |
| E. coli-derived recombinant human BMP-9 Ser320-Arg429 Accession # Q9UK05 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human and primate BMP-9 in ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human BMP-1, -1.1, -2, -3, -3b, -4, -5, -6, -7, -8, or -10 is observed. In direct ELISAs, approximately 80% cross-reactivity with recombinant mouse (rm) BMP-9 and 10% cross-reactivity with rmBMP-10 is observed. |
| 360107 | |
| Western Blot 1 ug/mL, Neutralization 0.01-0.04 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BMP9, BMP9BMP-9Bone morphogenetic protein 9, GDF2, GDF-2, growth differentiation factor 2, growth/differentiation factor 2 | |
| GDF2 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title