missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse PLC-delta3 Antibody, R&D Systems™

Description
PLC-delta 3 Polyclonal specifically detects PLC-delta 3 in Human, Mouse samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | PLC-delta 3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL |
| Gene Accession No. | Q8N3E9 |
| Gene Alias | EC 3.1.4.11, KIAA1964, MGC71172, Phosphoinositide phospholipase C-delta-3, phospholipase C, delta 3,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase delta-3, Phospholipase C-delta-3, PLC delta3, PLCD3, PLCdelta 3, PLC-delta-3 |
| Gene Symbols | PLCD3 |
| Host Species | Goat |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?