missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse PHOX2B Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF4940
This item is not returnable.
View return policy
Description
PHOX2B Polyclonal specifically detects PHOX2B in Human, Mouse samples. It is validated for Immunocytochemistry.
Specifications
| PHOX2B | |
| Polyclonal | |
| Unconjugated | |
| Q99453 | |
| PHOX2B | |
| E. coli-derived recombinant human PHOX2B Met1-Glu94 Accession # Q99453 | |
| 100 μg | |
| Primary | |
| Detects human PHOX2B in direct ELISAs. | |
| Human, Mouse | |
| IgG |
| Immunocytochemistry | |
| LYOPH | |
| Immunocytochemistry 5-15 ug/mL | |
| NBPHOX, NBPhoxPhox2b, Neuroblastoma Phox, paired mesoderm homeobox 2b, paired mesoderm homeobox protein 2B, paired-like homeobox 2bNBLST2, PHOX2B homeodomain protein, PMX2B, PMX2Bneuroblastoma paired-type homeobox protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8929 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction