missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Phospho-Tie-2 (Y992) Antibody, R&D Systems™
Rabbit Polyclonal Antibody has been used in 18 publications
Brand: R&D Systems AF2720
This item is not returnable.
View return policy
Description
Tie-2 Polyclonal specifically detects Tie-2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| Tie-2 (p tyr992) | |
| Polyclonal | |
| Unconjugated | |
| angiopoietin-1 receptor, CD202B, CD202b antigen, EC 2.7.10, EC 2.7.10.1, hTIE2, p140 TEK, soluble TIE2 variant 1, soluble TIE2 variant 2, TEK, TEK tyrosine kinase, endothelial, Tie2, TIE-2, TIE2CD202b, Tunica interna endothelial cell kinase, Tyrosine-protein kinase receptor TEK, Tyrosine-protein kinase receptor TIE-2, venous malformations, multiple cutaneous and mucosal, VMCM, VMCM1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 7010 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.5 μg/mL, Immunohistochemistry 1-15 μg/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| TEK | |
| Phosphopeptide containing human Tie-2 Y992 site | |
| 50 μg | |
| Primary | |
| Detects human and mouse Tie-2 when phosphorylated at Y992 in Western blots. | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction