missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Phospho-MSK2/RSKB (S196) Antibody, R&D Systems™

Rabbit Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF189
This item is not returnable.
View return policy
Description
MSK2 Polyclonal specifically detects MSK2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| MSK2 (p ser196) | |
| Polyclonal | |
| Unconjugated | |
| ribosomal protein S6 kinase, 90kDa, polypeptide 4, RPS6KA4, RSKB | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 8986 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| RPS6KA4 | |
| Phosphopeptide containing human and mouse MSK2/RSKB S196 site | |
| 100 μg | |
| Primary | |
| Detects human and mouse MSK2/RSKB when phosphorylated at S196. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction