missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Phospho-MSK2/RSKB (S196) Antibody, R&D Systems™

Rabbit Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF189-SP
This item is not returnable.
View return policy
Description
MSK2 Polyclonal specifically detects MSK2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| MSK2 (p ser196) | |
| Polyclonal | |
| Unconjugated | |
| Supplied as a 0.2 μm filtered solution in PBS with Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| RPS6KA4 | |
| Phosphopeptide containing human and mouse MSK2/RSKB S196 site | |
| 25 μg | |
| Primary | |
| Detects human and mouse MSK2/RSKB when phosphorylated at S196. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| ribosomal protein S6 kinase, 90kDa, polypeptide 4, RPS6KA4, RSKB | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 8986 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction