missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Oct-3/4 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 24 publications
Brand: R&D Systems MAB1759-SP
This item is not returnable.
View return policy
Description
Oct-3/4 Monoclonal specifically detects Oct-3/4 in Human, Mouse samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| Oct-3/4 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| Q01860 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5460 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
| 240408 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| POU5F1 | |
| E. coli-derived recombinant human Oct-3/4 Met1-Asn265 (Met262Leu) Accession # Q01860 | |
| 25 μg | |
| Primary | |
| Detects human Oct-3/4 in Western blots and detects mouse Oct-3/4 in flow cytometry. | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction