missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Nodal Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1315
This item is not returnable.
View return policy
Description
Nodal Polyclonal specifically detects Nodal in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Nodal | |
| Polyclonal | |
| Unconjugated | |
| P43021 | |
| NODAL | |
| E. coli-derived recombinant mouse Nodal His245-Leu354 Accession # P43021 | |
| 100 μg | |
| Primary | |
| Detects human and mouse Nodal in direct ELISAs and Western blots. | |
| Human, Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| BMP-16, MGC138230, nodal homolog, nodal homolog (mouse), nodal, mouse, homolog | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4838 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction