missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Neurturin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF477-SP
This item is not returnable.
View return policy
Description
Neurturin Polyclonal specifically detects Neurturin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction.
Specifications
| Neurturin | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| NRTN | |
| E. coli-derived recombinant mouse Neurturin | |
| 25 μg | |
| Primary | |
| Detects human and mouse Neurturin in direct ELISAs and Western blots. | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction 2.0 ug/mL | |
| neurturin, NRTN, NTN | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4902 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction