missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Myocardin Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB4028
This item is not returnable.
View return policy
Description
Myocardin Monoclonal specifically detects Myocardin in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Myocardin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q8IZQ8 | |
| MYOCD | |
| E. coli-derived recombinant human Myocardin Met97-Leu290 Accession # Q8IZQ8 | |
| 100 μg | |
| Primary | |
| Detects human and mouse Myocardin in Western blots. | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 355521 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BSAC2A, MYCD, myocardin, MYOCD, Srfcp | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 93649 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction