missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse MMP-24/MT5-MMP Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB924
This item is not returnable.
View return policy
Description
MMP-24/MT5-MMP Monoclonal specifically detects MMP-24/MT5-MMP in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| MMP-24/MT5-MMP | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.24, EC 3.4.24.-, EC 3.4.24.80, matrix metallopeptidase 24 (membrane-inserted), matrix metalloproteinase 24 (membrane-inserted), matrix metalloproteinase-24, membrane-type 5 matrix metalloproteinase, Membrane-type matrix metalloproteinase 5, Membrane-type-5 matrix metalloproteinase, MMP24, MMP-24, MT5MMP, MT5-MMP, MT5-MMPMMP25, MT-MMP 5, MTMMP5, MT-MMP5 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10893 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Flow Cytometry, CyTOF | |
| 143908 | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q9R0S2 | |
| MMP24 | |
| E. coli-derived recombinant mouse MMP-24/MT5-MMP Arg125-Gln296 Accession # Q9R0S2 | |
| 500 μg | |
| Primary | |
| Detects mouse MMP-24/MT5-MMP in direct ELISAs and detects human and mouse MMP-24/MT5-MMP in Western blots. In Western blots, less than 5% cross-reactivity with recombinant mouse (rm) MMP-3, rmMMP-9, and the catalytic domains of recombinant human MMP-14, 15, and 16 is observed. | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction