missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse MCPIP1 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 5 publications
Brand: R&D Systems MAB7875
This item is not returnable.
View return policy
Description
MCPIP1/ZC3H12A Monoclonal specifically detects MCPIP1/ZC3H12A in Human, Mouse samples. It is validated for Western Blot.
Specifications
| MCPIP1/ZC3H12A | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q5D1E8 | |
| ZC3H12A | |
| E. coli-derived recombinant human MCPIP1 Asp426-Glu599 Accession # Q5D1E8 | |
| 100 μg | |
| Primary | |
| Detects human MCPIP1 in direct ELISAs and human and mouse MCPIP1 in Western blots. In direct ELISAs, no cross-reactivity with recombinant human MCPIP3 is observed. | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 604421 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Reg1, Regnase-1, ZC3H12A, zinc finger CCCH-type containing 12A | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 80149 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction