missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Lysine (K)-specific Demethylase 4A/KDM4A Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF6434
This item is not returnable.
View return policy
Description
Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A Polyclonal specifically detects Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A | |
| Polyclonal | |
| Unconjugated | |
| AAH02558 | |
| KDM4A | |
| E. coli-derived recombinant human KDM4A Lys386-Ser514 Accession # AAH02558 | |
| 100 μg | |
| Primary | |
| Detects human KDM4A in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human KDM4C is observed. | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| EC 1.14.11, EC 1.14.11.-, JHDM3A, JHDM3AJMJD2, JmjC domain-containing histone demethylation protein 3A, JMJD2, JMJD2A, JMJD2Alysine-specific demethylase 4A, jumonji domain containing 2, jumonji domain containing 2A, Jumonji domain-containing protein 2A, KDM4A, KIAA0677jumonji C domain-containing histone demethylase 3A, lysine (K)-specific demethylase 4A, Lysine (K)specific Demethylase 4A | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 9682 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction