missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Lumican Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB28461
This item is not returnable.
View return policy
Description
Lumican Monoclonal specifically detects Lumican in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Lumican | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| NP_002336 | |
| LUM | |
| Mouse myeloma cell line NS0-derived recombinant human Lumican Gln19-Asn338 Accession # NP_002336 | |
| 100 μg | |
| Primary | |
| Detects human and mouse Lumican in direct ELISAs and Western blots. | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 669401 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| KSPG lumican, LDC, LUM, lumican, lumican proteoglycan, SLRR2D, SLRR2DLDCKeratan sulfate proteoglycan lumican | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4060 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction