missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse LRRTM3 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF4898
This item is not returnable.
View return policy
Description
LRRTM3 Polyclonal specifically detects LRRTM3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| LRRTM3 | |
| Polyclonal | |
| Unconjugated | |
| Q86VH5 | |
| LRRTM3 | |
| Mouse myeloma cell line NS0-derived recombinant human LRRTM3 Glu31-Lys419 Accession # Q86VH5 | |
| 100 μg | |
| Primary | |
| Detects human and mouse LRRTM3 in direct ELISAs and Western blots. In direct ELISA, approximately 25% cross-reactivity with recombinant human (rh) LRRTM4, and less than 5% cross-reactivity with rhLRRTM1 and rhLRRTM2 is observed. | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| leucine rich repeat transmembrane neuronal 3, leucine-rich repeat transmembrane neuronal protein 3, MGC131810 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 347731 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction